Anti-IgE Goat Polyclonal Antibody (SureLight® APC (Allophycocyanin))

Lieferant: Abcam
AB99904-100
ABCAAB99904-100EA 676 EUR
ABCAAB99904-100
Anti-IgE Goat Polyclonal Antibody (SureLight® APC (Allophycocyanin))
Antikörper
Type: Secondary
Antigen: IgE (Mouse) (mouse IgE)
Clonality: polyclonal
Clone:
Conjugation: SureLight® Allophycocyanin
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR