Anti-IgE Goat Polyclonal Antibody (SureLight® PE (Phycoerytrhin))

Lieferant: Abcam
AB99910-100
ABCAAB99910-100EA 840 EUR
ABCAAB99910-100
Anti-IgE Goat Polyclonal Antibody (SureLight® PE (Phycoerytrhin))
Antikörper
Type: Secondary
Antigen: IgE
Clonality: Polyclonal
Clone:
Conjugation: SureLight® PE
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR