Anti-IgM Goat Polyclonal Antibody (SureLight® PE (Phycoerytrhin))

Lieferant: Abcam
AB99917-100
ABCAAB99917-100EA 676 EUR
ABCAAB99917-100
Anti-IgM Goat Polyclonal Antibody (SureLight® PE (Phycoerytrhin))
Antikörper
Type: Secondary
Antigen: IgM (Mouse) (mouse IgM)
Clonality: polyclonal
Clone:
Conjugation: SureLight® PE
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR