Anti-SUMO3 Mouse Monoclonal Antibody

Lieferant: Abgent
AM1201A
ABGEAM1201AEA 354 EUR
ABGEAM1201A
Anti-SUMO3 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: SUMO3 (small ubiquitin-like modifier 3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR