Anti-MMP2 Mouse Monoclonal Antibody

Lieferant: Abgent
AM1844A
ABGEAM1844AEA 354 EUR
ABGEAM1844A
Anti-MMP2 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: MMP2 (matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR