Anti-THAP11 Mouse Monoclonal Antibody

Lieferant: Abgent
AM1984B
ABGEAM1984BEA 354 EUR
ABGEAM1984B
Anti-THAP11 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: THAP11 (THAP domain containing 11)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Mouse
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR