Anti-BTK Mouse Monoclonal Antibody [clone: 488CT2.1.2]

Lieferant: Abgent
AM2062A
ABGEAM2062AEA 354 EUR
ABGEAM2062A
Anti-BTK Mouse Monoclonal Antibody [clone: 488CT2.1.2]
Antikörper
Type: Primary
Antigen: BTK
Clonality: Monoclonal
Clone: 488CT2.1.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR