Anti-IL1B Mouse Monoclonal Antibody [clone: 614CT4.3.1]

Lieferant: Abgent
AM2121A
ABGEAM2121AEA 354 EUR
ABGEAM2121A
Anti-IL1B Mouse Monoclonal Antibody [clone: 614CT4.3.1]
Antikörper
Type: Primary
Antigen: IL1B
Clonality: Monoclonal
Clone: 614CT4.3.1
Conjugation: Unconjugated
Epitope: internal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR