Anti-TECRL Mouse Monoclonal Antibody [clone: 601CT22.4.3]

Lieferant: Abgent
AM2127A
ABGEAM2127AEA 354 EUR
ABGEAM2127A
Anti-TECRL Mouse Monoclonal Antibody [clone: 601CT22.4.3]
Antikörper
Type: Primary
Antigen: TECRL
Clonality: Monoclonal
Clone: 601CT22.4.3
Conjugation: Unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR