Anti-PLP1 Mouse Monoclonal Antibody [clone: 538CT16.5.4]
Lieferant: Abgent
AM2143B
ABGEAM2143BEA 354 EUR
ABGEAM2143B
Anti-PLP1 Mouse Monoclonal Antibody [clone: 538CT16.5.4]
Antikörper
Type: Primary
Antigen: PLP1
Clonality: Monoclonal
Clone: 538CT16.5.4
Conjugation: Unconjugated
Epitope: internal
Host: Mouse
Isotype: IgM
Reactivity: Human