Anti-AR Mouse Monoclonal Antibody [clone: 549CT16.1.4]

Lieferant: Abgent
AM2145B
ABGEAM2145BEA 354 EUR
ABGEAM2145B
Anti-AR Mouse Monoclonal Antibody [clone: 549CT16.1.4]
Antikörper
Type: Primary
Antigen: AR
Clonality: Monoclonal
Clone: 549CT16.1.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgA
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR