Anti-SOCS3 Mouse Monoclonal Antibody [clone: 532CT13.5.3]

Lieferant: Abgent
AM2154A
ABGEAM2154AEA 354 EUR
ABGEAM2154A
Anti-SOCS3 Mouse Monoclonal Antibody [clone: 532CT13.5.3]
Antikörper
Type: Primary
Antigen: SOCS3
Clonality: Monoclonal
Clone: 532CT13.5.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR