Anti-KLF4 Mouse Monoclonal Antibody

Lieferant: Abgent
AM2725A
ABGEAM2725AEA 354 EUR
ABGEAM2725A
Anti-KLF4 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: KLF4 (Kruppel-like factor 4 (gut))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR