Anti-AKR1C3 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP10158A
ABGEAP10158AEA 354 EUR
ABGEAP10158A
Anti-AKR1C3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AKR1C3 (aldo-keto reductase family 1, member C3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR