Anti-CCL4 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP10172B
ABGEAP10172BEA 354 EUR
ABGEAP10172B
Anti-CCL4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CCL4 (chemokine (C-C motif) ligand 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse