Anti-FUS Rabbit Polyclonal Antibody
Lieferant: Abgent
AP10187B
ABGEAP10187BEA 354 EUR
ABGEAP10187B
Anti-FUS Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FUS (fused in sarcoma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse