Anti-Dnmt3A/Dnmt3A2 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP1023B
ABGEAP1023BEA 354 EUR
ABGEAP1023B
Anti-Dnmt3A/Dnmt3A2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Dnmt3A/Dnmt3A2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR