Anti-CALML4 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP10243B
ABGEAP10243BEA 354 EUR
ABGEAP10243B
Anti-CALML4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CALML4 (calmodulin-like 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR