Anti-CCDC148 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP10609C
ABGEAP10609CEA 354 EUR
ABGEAP10609C
Anti-CCDC148 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CCDC148 (coiled-coil domain containing 148)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR