Anti-DCAF10 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP10636B
ABGEAP10636BEA 354 EUR
ABGEAP10636B
Anti-DCAF10 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DCAF10 (DDB1 and CUL4 associated factor 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human