Anti-UBA2 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP1065B
ABGEAP1065BEA 354 EUR
ABGEAP1065B
Anti-UBA2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: UBA2 (ubiquitin-like modifier activating enzyme 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR