Anti-APCS Rabbit Polyclonal Antibody

Lieferant: Abgent
AP10677B
ABGEAP10677BEA 354 EUR
ABGEAP10677B
Anti-APCS Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: APCS (amyloid P component, serum)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR