Anti-COL18A1 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP10706C
ABGEAP10706CEA 354 EUR
ABGEAP10706C
Anti-COL18A1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: COL18A1 (collagen, type XVIII, alpha 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR