Anti-ATP8B2 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP10779A
ABGEAP10779AEA 354 EUR
ABGEAP10779A
Anti-ATP8B2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ATP8B2 (ATPase, aminophospholipid transporter, class I, type 8B, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human