Anti-LCE1A Rabbit Polyclonal Antibody
Lieferant: Abgent
AP10891B
ABGEAP10891BEA 354 EUR
ABGEAP10891B
Anti-LCE1A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LCE1A (late cornified envelope 1A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human