Anti-NFE2L1 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP10927B
ABGEAP10927BEA 354 EUR
ABGEAP10927B
Anti-NFE2L1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NFE2L1 (nuclear factor, erythroid 2-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR