Anti-OR5L2 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP11066A
ABGEAP11066AEA 354 EUR
ABGEAP11066A
Anti-OR5L2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: OR5L2 (olfactory receptor, family 5, subfamily L, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human