Anti-GAD2 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP11118C
ABGEAP11118CEA 354 EUR
ABGEAP11118C
Anti-GAD2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GAD2 (glutamate decarboxylase 2 (pancreatic islets and brain, 65kDa))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human