Anti-FGL2 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP11121B
ABGEAP11121BEA 354 EUR
ABGEAP11121B
Anti-FGL2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FGL2 (fibrinogen-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR