Anti-CUZD1 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP11139B
ABGEAP11139BEA 354 EUR
ABGEAP11139B
Anti-CUZD1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CUZD1 (CUB and zona pellucida-like domains 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR