Anti-PPP1R21 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP11169C
ABGEAP11169CEA 354 EUR
ABGEAP11169C
Anti-PPP1R21 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PPP1R21 (protein phosphatase 1, regulatory subunit 21)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR