Anti-EPGN Rabbit Polyclonal Antibody [clone: 143CT50.11.8]

Lieferant: Abgent
AP11242A
ABGEAP11242AEA 354 EUR
ABGEAP11242A
Anti-EPGN Rabbit Polyclonal Antibody [clone: 143CT50.11.8]
Antikörper
Type: Primary
Antigen: EPGN (epithelial mitogen)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR