Anti-OR4F15 Rabbit Polyclonal Antibody [clone: 199CT6.1.2]
Lieferant: Abgent
AP11344A
ABGEAP11344AEA 354 EUR
ABGEAP11344A
Anti-OR4F15 Rabbit Polyclonal Antibody [clone: 199CT6.1.2]
Antikörper
Type: Primary
Antigen: OR4F15 (olfactory receptor, family 4, subfamily F, member 15)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human