Anti-CPM Rabbit Polyclonal Antibody [clone: 57CT23.3.4]
Lieferant: Abgent
AP11458C
ABGEAP11458CEA 354 EUR
ABGEAP11458C
Anti-CPM Rabbit Polyclonal Antibody [clone: 57CT23.3.4]
Antikörper
Type: Primary
Antigen: CPM (carboxypeptidase M)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human