Anti-CCDC54 Rabbit Polyclonal Antibody [clone: 46CT61.6.4]

Lieferant: Abgent
AP11474C
ABGEAP11474CEA 354 EUR
ABGEAP11474C
Anti-CCDC54 Rabbit Polyclonal Antibody [clone: 46CT61.6.4]
Antikörper
Type: Primary
Antigen: CCDC54 (coiled-coil domain containing 54)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR