Anti-MPO Rabbit Polyclonal Antibody
Lieferant: Abgent
AP11560A
ABGEAP11560AEA 354 EUR
ABGEAP11560A
Anti-MPO Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MPO (myeloperoxidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human