Anti-AKR1C3 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP11656C
ABGEAP11656CEA 354 EUR
ABGEAP11656C
Anti-AKR1C3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AKR1C3 (aldo-keto reductase family 1, member C3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human