Anti-BTC Rabbit Polyclonal Antibody

Lieferant: Abgent
AP11669A
ABGEAP11669AEA 354 EUR
ABGEAP11669A
Anti-BTC Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BTC (betacellulin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR