Anti-LOC129293 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP11741B
ABGEAP11741BEA 354 EUR
ABGEAP11741B
Anti-LOC129293 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LOC129293
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR