Anti-SET7 (SET9) Rabbit Polyclonal Antibody

Lieferant: Abgent
AP1194C
ABGEAP1194CEA 354 EUR
ABGEAP1194C
Anti-SET7 (SET9) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SET7 (SET9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR