Anti-METTL2B Rabbit Polyclonal Antibody

Lieferant: Abgent
AP11952B
ABGEAP11952BEA 354 EUR
ABGEAP11952B
Anti-METTL2B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: METTL2B (methyltransferase like 2B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR