Anti-FOLR4 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP11990A
ABGEAP11990AEA 354 EUR
ABGEAP11990A
Anti-FOLR4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FOLR4 (folate receptor 4, delta (putative))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human