Anti-OR4L1 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP12134B
ABGEAP12134BEA 354 EUR
ABGEAP12134B
Anti-OR4L1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: OR4L1 (olfactory receptor, family 4, subfamily L, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR