Anti-TAF4B Rabbit Polyclonal Antibody

Lieferant: Abgent
AP12163B
ABGEAP12163BEA 354 EUR
ABGEAP12163B
Anti-TAF4B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TAF4B (TAF4b RNA polymerase II, TATA box binding protein (TBP)-associated factor, 105kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR