Anti-COX7A2L Rabbit Polyclonal Antibody
Lieferant: Abgent
AP12338C
ABGEAP12338CEA 354 EUR
ABGEAP12338C
Anti-COX7A2L Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: COX7A2L (cytochrome c oxidase subunit VIIa polypeptide 2 like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse