Anti-FOXI2 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP12339C
ABGEAP12339CEA 354 EUR
ABGEAP12339C
Anti-FOXI2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FOXI2 (forkhead box I2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR