Anti-TSSK6 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP12462A
ABGEAP12462AEA 354 EUR
ABGEAP12462A
Anti-TSSK6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TSSK6 (testis-specific serine kinase 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human