Anti-MINA Rabbit Polyclonal Antibody
Lieferant: Abgent
AP12466A
ABGEAP12466AEA 354 EUR
ABGEAP12466A
Anti-MINA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MINA (MYC induced nuclear antigen)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human