Anti-RNF11 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP12908C
ABGEAP12908CEA 354 EUR
ABGEAP12908C
Anti-RNF11 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RNF11 (ring finger protein 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR