Anti-BID Rabbit Polyclonal Antibody

Lieferant: Abgent
AP1307A
ABGEAP1307AEA 354 EUR
ABGEAP1307A
Anti-BID Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BID (BH3 interacting domain death agonist)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: BH3 domain
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR